6-Fluoro-o-toluonitrile 97%

Supplier: Ambeed
Warning

A101622-100G A101622-10G A101622-25G A101622-1G A101622-5G
103796-052EA 10 USD
103796-052 103821-744 77107-208 77107-210 77107-212 77297-170 77297-172
6-Fluoro-o-toluonitrile 97%
6-Fluoro-o-toluonitrile

Precaución: for Research Use Only

Formula: C₈H₆FN
MW: 135.14 g/mol
Boiling Pt: 207.4 °C (760 mm Hg)
Melting Pt: 39…46 °C
Flash Pt: 90.5 °C
Storage Temperature: Ambient
MDL Number: MFCD06660277
CAS Number: 198633-76-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR