6-Fluoro-o-toluidine 97%

Supplier: Ambeed
Warning

Synonyms: 2-Fluoro-6-methylaniline

A486647-10G A486647-25G A486647-5G A486647-1G A486647-100G
103828-182EA 9 USD
103828-182 103839-256 76774-418 76843-206 76843-208 77307-566 77626-970
6-Fluoro-o-toluidine 97%
6-Fluoro-o-toluidine

Precaución: for Research Use Only

Formula: C₇H₈FN
MW: 125.1435 g/mol
Boiling Pt: 187 °C (760 mm Hg)
Flash Pt: 80.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06658252
CAS Number: 443-89-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR