2-Fluoro-6-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine ≥95%

Supplier: Ambeed
Warning

A247568-100MG A247568-250MG A247568-5G A247568-1G
103811-782EA 173 USD
103811-782 76987-810 76987-812 76987-814 77090-960
2-Fluoro-6-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine ≥95%
2-Fluoro-6-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

Precaución: for Research Use Only

Formula: C₁₂H₁₇BFNO₂
MW: 237.08 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08063098
CAS Number: 1310384-07-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR