2-Fluoro-3-nitro-6-picoline 97%

Supplier: Ambeed
Warning

Synonyms: 2-Fluoro-3-nitro-6-methylpyridine, 2-Fluoro-6-methyl-3-nitropyridine

A195251-1G A195251-250MG A195251-10G
103816-732EA 17 USD
103816-732 103847-346 77085-298 77097-218 77685-483
2-Fluoro-3-nitro-6-picoline 97%
2-Fluoro-3-nitro-6-picoline

Precaución: for Research Use Only

Formula: C₆H₅FN₂O₂
MW: 156.12 g/mol
Boiling Pt: 266 °C (760 mm Hg)
Flash Pt: 114.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03095067
CAS Number: 19346-45-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR