2-Fluoro-6-methoxyphenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-Fluoro-6-methoxybenzeneboronic acid

A213302-1G A213302-25G A213302-5G A213302-100G A213302-10G A213302-500G
103818-286EA 5 USD
103818-286 103848-992 77087-494 77087-496 77087-498 77192-694 77192-696 77192-698
2-Fluoro-6-methoxyphenylboronic acid 98%
2-Fluoro-6-methoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₈BFO₃
MW: 169.95 g/mol
Boiling Pt: 303.1 °C (760 mm Hg)
Melting Pt: 120…125 °C
Flash Pt: 137.1 °C
Storage Temperature: Ambient
MDL Number: MFCD02179483
CAS Number: 78495-63-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR