2-Fluoro-6-methoxyphenol 98%

Supplier: Ambeed
Warning

A674419-1G A674419-25G A674419-5G A674419-250MG
103840-514EA 37 USD
103840-514 76844-708 76844-710 76844-712 77267-294
2-Fluoro-6-methoxyphenol 98%
2-Fluoro-6-methoxyphenol

Precaución: for Research Use Only

Formula: C₇H₇FO₂
MW: 142.13 g/mol
Boiling Pt: 224 °C (760 mm Hg)
Flash Pt: 103.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00075140
CAS Number: 73943-41-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR