2-Fluoro-6-methoxybenzoic acid 97%
Supplier: Ambeed
|
Warning
|
A100559-100G
A100559-10G
A100559-25G
A100559-1G
A100559-5G
103795-510EA
9
USD
103795-510
103821-200
76994-450
76994-452
76994-454
77282-510
77296-756
2-Fluoro-6-methoxybenzoic acid 97%
2-Fluoro-6-methoxybenzoic acid
Precaución: for Research Use Only
Formula:
C₈H₇FO₃ Flash Pt: 122.9 °C Storage Temperature: Ambient |
MDL Number:
MFCD00671764 CAS Number: 137654-21-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...