6-Fluoro-o-anisaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: 2-Fluoro-6-mehoxybenzaldehyde, 2-Fluoro-6-methoxybenzaldehyde, 6-Fluoro-2-anisaldehyde

A111603-10G A111603-250MG A111603-1G A111603-5G
103797-870EA 14 USD
103797-870 103823-688 77233-598 77233-600 77310-502 77310-504
6-Fluoro-o-anisaldehyde 98%
6-Fluoro-o-anisaldehyde

Precaución: for Research Use Only

Formula: C₈H₇FO₂
MW: 154.14 g/mol
Melting Pt: 59…63 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD01090998
CAS Number: 146137-74-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR