2-Fluoro-6-methoxyaniline 98%

Supplier: Ambeed
Warning

A237751-1G A237751-25G A237751-5G A237751-100MG A237751-10G
103811-176EA 8 USD
103811-176 103850-958 77090-056 77090-058 77090-060 77261-698 77261-700
2-Fluoro-6-methoxyaniline 98%
2-Fluoro-6-methoxyaniline

Precaución: for Research Use Only

Formula: C₇H₈FNO
MW: 141.1429 g/mol
Boiling Pt: 189.2 °C (760 mm Hg)
Flash Pt: 68.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03788538
CAS Number: 446-61-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR