2-Fluoro-6-iodobenzonitrile 98%

Supplier: Ambeed
Warning

A163290-100G A163290-10G
103843-968EA 9 USD
103843-968 76768-704 77877-075
2-Fluoro-6-iodobenzonitrile 98%
2-Fluoro-6-iodobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₃FIN
Melting Pt: 49…53 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00015478
CAS Number: 79544-29-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR