2-Fluoro-6-hydroxybenzonitrile 98%

Supplier: Ambeed
Warning

A651284-100G A651284-10G A651284-250MG A651284-1G A651284-25G A651284-5G
103840-398EA 15 USD
103840-398 76775-556 76775-558 76775-560 76844-564 76844-566 76844-568
2-Fluoro-6-hydroxybenzonitrile 98%
2-Fluoro-6-hydroxybenzonitrile

Precaución: for Research Use Only

Formula: C₇H₄FNO
MW: 137.11 g/mol
Melting Pt: 154…158 °C
Flash Pt: 109.5 °C
Storage Temperature: Ambient
MDL Number: MFCD03428592
CAS Number: 140675-43-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR