6-Fluorosalicylic acid 98%

Supplier: Ambeed
Warning

Synonyms: 6-Fluoro-2-hydroxybenzoic acid, 2-Fluoro-6-hydroxybenzoic acid

A182998-100G A182998-10G A182998-1G A182998-25G A182998-5G
103815-590EA 9 USD
103815-590 103846-152 76769-616 76769-618 76769-620 77083-734 77083-736 77083-738
6-Fluorosalicylic acid 98%
6-Fluorosalicylic acid

Precaución: for Research Use Only

Formula: C₇H₅FO₃
Boiling Pt: 291.2 °C (760 mm Hg)
Melting Pt: 159…163 °C
Flash Pt: 80.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00052179
CAS Number: 67531-86-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR