6-Fluorosalicylaldehyde 97%
Supplier: Ambeed
|
Warning
|
A277942-1G
A277942-5G
A277942-10G
103813-384EA
11
USD
103813-384
103853-322
77093-226
77093-228
77141-120
6-Fluorosalicylaldehyde 97%
6-Fluorosalicylaldehyde
Precaución: for Research Use Only
Formula:
C₇H₅FO₂ Boiling Pt: 201.3 °C (760 mm Hg) Melting Pt: 37…39 °C Flash Pt: 75.6 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD01090996 CAS Number: 38226-10-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...