2-Fluoro-6-(trifluoromethyl)benzonitrile 97%

Supplier: Ambeed
Warning

Synonyms: 2-Cyano-3-fluorobenzotrifluoride

A380007-1G A380007-25G A380007-100G A380007-10G
103826-660EA 5 USD
103826-660 103837-696 76841-018 76841-020 77195-626 77195-628
2-Fluoro-6-(trifluoromethyl)benzonitrile 97%
2-Fluoro-6-(trifluoromethyl)benzonitrile

Precaución: for Research Use Only

Formula: C₈H₃F₄N
MW: 189.11 g/mol
Flash Pt: 99 °C
Storage Temperature: Ambient
MDL Number: MFCD00061153
CAS Number: 133116-83-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR