2-Fluoro-6-(trifluoromethyl)aniline 98%

Supplier: Ambeed
Danger

Synonyms: 2-Amino-α,α,α,3-tetrafluorotoluene, 2-Amino-alpha,alpha,alpha,3-tetrafluorotoluene, 2-Amino-3-fluorobenzotrifluoride, α,α,α,6-Tetrafluoro-o-toluidine

A257795-1G A257795-25G A257795-10G
103812-338EA 10 USD
103812-338 103852-206 77091-730 77091-732 77140-666
2-Fluoro-6-(trifluoromethyl)aniline 98%
2-Fluoro-6-(trifluoromethyl)aniline

Precaución: for Research Use Only

Formula: C₇H₅F₄N
MW: 179.12
Boiling Pt: 155
Density: 1.388
Flash Pt: 53 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00061242
CAS Number: 144851-61-6
UN: 1993
ADR: 3 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR