2-Fluoro-5-pyridineboronic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
6-Fluoropyridine-3-boronic acid, 2-Fluoropyridine-5-boronic acid, 2-Fluoro-5-pyridylboronic acid, 2-Fluoro-5-pyridinylboronic acid, 6-Fluoro-3-pyridinylboronic acid
A142326-100G
A142326-10G
A142326-250MG
A142326-500G
A142326-1G
A142326-25G
A142326-5G
103803-144EA
6
USD
103803-144
77010-396
77010-398
77010-400
77010-402
77316-360
77316-362
77316-364
2-Fluoro-5-pyridineboronic acid 98%
2-Fluoro-5-pyridineboronic acid
Precaución: for Research Use Only
Formula:
C₅H₅BFNO₂ MW: 140.91 g/mol Boiling Pt: 326.8 °C (760 mm Hg) Flash Pt: 142.2 °C Storage Temperature: Ambient |
MDL Number:
MFCD03411559 CAS Number: 351019-18-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...