2-Fluoro-5-nitropyridine 97%

Supplier: Ambeed
Warning

A254421-100G A254421-10G A254421-5G A254421-25G A254421-500G
103812-134EA 5 USD
103812-134 103851-988 76976-756 76976-758 77091-430 77304-630 77672-274
2-Fluoro-5-nitropyridine 97%
2-Fluoro-5-nitropyridine

Precaución: for Research Use Only

Formula: C₅H₃FN₂O₂
Boiling Pt: 237.5 °C (760 mm Hg)
Melting Pt: 142…144 °C
Flash Pt: 97.5 °C
Storage Temperature: Ambient
MDL Number: MFCD03095059
CAS Number: 456-24-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR