2-Fluoro-5-nitrobenzaldehyde 98%

Supplier: Ambeed
Warning

A104830-100G A104830-10G A104830-25G A104830-1G A104830-5G
103796-784EA 7 USD
103796-784 103822-510 76765-434 76765-436 76765-438 77283-268 77309-368
2-Fluoro-5-nitrobenzaldehyde 98%
2-Fluoro-5-nitrobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄FNO₃
MW: 169.11 g/mol
Boiling Pt: 269.7 °C (760 mm Hg)
Melting Pt: 57…60 °C
Flash Pt: 116.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00042298
CAS Number: 27996-87-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR