2-Fluoro-5-nitroaniline 98%

Supplier: Ambeed
Warning

A123106-100G A123106-10G A123106-25G
103808-264EA 374 USD
103808-264 77284-950 77284-952 77284-954
2-Fluoro-5-nitroaniline 98%
2-Fluoro-5-nitroaniline

Precaución: for Research Use Only

Formula: C₆H₅FN₂O₂
MW: 156.12 g/mol
Boiling Pt: 295.5 °C (760 mm Hg)
Melting Pt: 97…100 °C
Flash Pt: 91 °C
Storage Temperature: Ambient
MDL Number: MFCD00007652
CAS Number: 369-36-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR