6-Fluoro-m-cresol 98%

Supplier: Ambeed
Warning

Synonyms: 2-Fluoro-5-methylphenol, 1-Fluoro-2-hydroxy-4-methylbenzene

A143677-100G A143677-1G A143677-25G
103803-308EA 11 USD
103803-308 76767-924 77316-600 77316-602
6-Fluoro-m-cresol 98%
6-Fluoro-m-cresol

Precaución: for Research Use Only

Formula: C₇H₇FO
MW: 126.13 g/mol
Boiling Pt: 173 °C (760 mm Hg)
Flash Pt: 67 °C
Storage Temperature: Ambient
MDL Number: MFCD00190101
CAS Number: 63762-79-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR