2-Fluoro-5-methylbenzaldehyde 98%

Supplier: Ambeed
Warning

A279282-100G A279282-10G A279282-1G A279282-5G A279282-25G
103813-460EA 5 USD
103813-460 103853-404 76692-992 76692-994 77093-340 77093-342 77305-322
2-Fluoro-5-methylbenzaldehyde 98%
2-Fluoro-5-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₇FO
Boiling Pt: 198.2 °C (760 mm Hg)
Flash Pt: 80 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02093762
CAS Number: 93249-44-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR