2-Fluoro-5-methoxyaniline 97%

Supplier: Ambeed
Warning

Synonyms: 3-Amino-4-fluoroanisole, 6-Fluoro-m-anisidine

A484393-10G A484393-1G
103828-162EA 18 USD
103828-162 103839-236 76789-704 76843-174
2-Fluoro-5-methoxyaniline 97%
2-Fluoro-5-methoxyaniline

Precaución: for Research Use Only

Formula: C₇H₈FNO
MW: 141.143 g/mol
Boiling Pt: 237.6 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD11226283
CAS Number: 62257-15-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR