2-Fluoro-5-iodopyridine 98%

Supplier: Ambeed
Warning

A153880-100G A153880-10G A153880-500G A153880-1G A153880-25G A153880-5G
103804-574EA 5 USD
103804-574 103842-752 76984-746 76984-748 76984-750 77300-032 77318-434 77318-436
2-Fluoro-5-iodopyridine 98%
2-Fluoro-5-iodopyridine

Precaución: for Research Use Only

Formula: C₅H₃FIN
MW: 222.99 g/mol
Flash Pt: 101 °C
Storage Temperature: Ambient
MDL Number: MFCD03095301
CAS Number: 171197-80-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR