2-Fluoro-5-iodobenzoic acid 98%

Supplier: Ambeed
Warning

A226632-1G A226632-25G A226632-100G A226632-10G
103819-256EA 5 USD
103819-256 103850-160 77088-894 77088-896 77139-944 77139-946
2-Fluoro-5-iodobenzoic acid 98%
2-Fluoro-5-iodobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₄FIO₂
MW: 266.01 g/mol
Melting Pt: 164…168 °C
Flash Pt: 150.2 °C
Storage Temperature: Ambient
MDL Number: MFCD03094517
CAS Number: 124700-41-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR