2-Fluoro-5-hydroxybenzenecarbonitrile 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Fluoro-5-hydroxybenzonitrile
A214591-100MG
A214591-10G
A214591-250MG
A214591-1G
A214591-25G
A214591-5G
103818-416EA
11
USD
103818-416
103849-120
77030-610
77030-612
77030-614
77087-646
77087-648
77087-650
2-Fluoro-5-hydroxybenzenecarbonitrile 97%
2-Fluoro-5-hydroxybenzenecarbonitrile
Precaución: for Research Use Only
Formula:
C₇H₄FNO Boiling Pt: 268.2 °C (760 mm Hg) Melting Pt: 111…113 °C Flash Pt: 116 °C Storage Temperature: Ambient |
MDL Number:
MFCD11846514 CAS Number: 104798-53-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...