2-Fluoro-5-pyridineboronic acid pinacol ester 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-(6-Fluoro-3-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-Fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 6-Fluoropyridine-3-boronic acid Pinacol ester, 2-Fluoropyridine-5-boronic acid pinacol ester
A197554-1G
A197554-25G
A197554-5G
A197554-100G
A197554-10G
A197554-250MG
103816-936EA
11
USD
103816-936
103847-556
77085-568
77085-570
77085-572
77260-708
77260-710
77260-712
2-Fluoro-5-pyridineboronic acid pinacol ester 97%
2-Fluoro-5-pyridineboronic acid pinacol ester
Precaución: for Research Use Only
Formula:
C₁₁H₁₅BFNO₂ MW: 223.05 g/mol Boiling Pt: 304.2 °C (760 mm Hg) Melting Pt: 29…33 °C Flash Pt: 137 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD03412792 CAS Number: 444120-95-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...