3-Amino-4-fluorophenylboronic acid pinacol ester 97%

Supplier: Ambeed
Warning

Synonyms: 3-Amino-4-fluorobenzeneboronic acid pinacol ester, 2-Fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline, 3-Amino-4-fluorophenylboronic acid, pinacol ester

A212809-250MG A212809-10G
103818-238EA 31 USD
103818-238 103848-934 76873-730 77883-514
3-Amino-4-fluorophenylboronic acid pinacol ester 97%
3-Amino-4-fluorophenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₇BFNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD11044430
CAS Number: 1003575-43-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR