2-Fluoro-5-[(4-oxo-3,4-dihydrophthalazin-1-yl)methyl]benzoic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-Fluoro-5-((4-oxo-3,4-dihydrophthalazin-1-yl)methyl)benzoic acid

A330791-1G A330791-25G A330791-5G A330791-10G A330791-250MG A330791-100G
103825-562EA 8 USD
103825-562 103836-524 76839-506 76839-508 76839-510 76876-000 76876-002 77612-900
2-Fluoro-5-[(4-oxo-3,4-dihydrophthalazin-1-yl)methyl]benzoic acid 98%
2-Fluoro-5-[(4-oxo-3,4-dihydrophthalazin-1-yl)methyl]benzoic acid

Precaución: for Research Use Only

Formula: C₁₆H₁₁FN₂O₃
MW: 298.269 g/mol
Melting Pt: 260…264 °C
Storage Temperature: Ambient
MDL Number: MFCD14636678
CAS Number: 763114-26-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR