2-Fluoro-4-nitrobenzonitrile 98%

Supplier: Ambeed
Warning

A158701-100G A158701-25G A158701-1G
103805-160EA 9 USD
103805-160 103843-374 77110-572 77300-358 77319-232
2-Fluoro-4-nitrobenzonitrile 98%

Precaución: for Research Use Only

Formula: C₇H₃FN₂O₂
Melting Pt: 70…71 °C
Storage Temperature: Ambient
MDL Number: MFCD04038715
CAS Number: 34667-88-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR