2-Fluoro-p-toluic acid 98%
Supplier: Ambeed
|
Warning
|
A111555-100G
A111555-10G
A111555-25G
A111555-500G
A111555-1G
A111555-5G
103797-860EA
5
USD
103797-860
103823-678
77233-590
77233-592
77233-594
77233-596
77310-486
77310-488
2-Fluoro-p-toluic acid 98%
2-Fluoro-p-toluic acid
Precaución: for Research Use Only
Formula:
C₈H₇FO₂ Boiling Pt: 271.7 °C (760 mm Hg) Melting Pt: 186…189 °C Storage Temperature: Ambient |
MDL Number:
MFCD03092906 CAS Number: 7697-23-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...