2-Fluoro-4-methylbenzaldehyde 98%
Supplier: Ambeed
|
Warning
|
A109520-100G
A109520-10G
A109520-250MG
A109520-25G
A109520-5G
103797-518EA
9
USD
103797-518
103823-316
76687-424
76687-426
76687-428
76687-430
77310-136
2-Fluoro-4-methylbenzaldehyde 98%
2-Fluoro-4-methylbenzaldehyde
Precaución: for Research Use Only
Formula:
C₈H₇FO Boiling Pt: 202.1 °C (760 mm Hg) Flash Pt: 84.3 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD03094325 CAS Number: 146137-80-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...