2-Fluoro-4-methyl-5-pyridineboronic acid 97%

Supplier: Ambeed
Warning

A167917-100G A167917-100MG A167917-10G A167917-250MG A167917-25G A167917-5G
103806-228EA 15 USD
103806-228 103844-486 77028-894 77028-896 77028-898 77028-900 77081-518 77081-520
2-Fluoro-4-methyl-5-pyridineboronic acid 97%
2-Fluoro-4-methyl-5-pyridineboronic acid

Precaución: for Research Use Only

Formula: C₆H₇BFNO₂
Boiling Pt: 331.6 °C (760 mm Hg)
Flash Pt: 154.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08741406
CAS Number: 1072944-18-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR