(2-Fluoro-4-methoxyphenyl)acetic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2-Fluoro-4-methoxyphenylacetic acid

A268536-250MG A268536-1G
103812-920EA 44 USD
103812-920 103852-822 76875-006 77305-006
(2-Fluoro-4-methoxyphenyl)acetic acid 97%
(2-Fluoro-4-methoxyphenyl)acetic acid

Precaución: for Research Use Only

Formula: C₉H₉FO₃
MW: 184.17 g/mol
Boiling Pt: 290.6 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD06800744
CAS Number: 883531-28-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR