2-Fluoro-p-anisaldehyde 95%

Supplier: Ambeed
Warning

Synonyms: 2-Fluoro-4-methoxybenzaldehyde, 2-Fluoro-4-anisaldehyde

A299708-100G A299708-10G A299708-25G A299708-1G A299708-5G
103814-236EA 6 USD
103814-236 103835-386 76693-388 76693-390 76838-078 77305-778 77305-780
2-Fluoro-p-anisaldehyde 95%
2-Fluoro-p-anisaldehyde

Precaución: for Research Use Only

Formula: C₈H₇FO₂
Melting Pt: 43…48 °C
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00236679
CAS Number: 331-64-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR