2-Fluoro-4-iodopyridine 98%

Supplier: Ambeed
Warning

A211477-100G A211477-10G A211477-500G A211477-1G A211477-25G A211477-5G
103818-110EA 5 USD
103818-110 103848-804 77030-510 77030-512 77030-514 77087-210 77087-212 77087-214
2-Fluoro-4-iodopyridine 98%
2-Fluoro-4-iodopyridine

Precaución: for Research Use Only

Formula: C₅H₃FIN
MW: 222.99 g/mol
Boiling Pt: 223 °C (760 mm Hg)
Melting Pt: 57…58 °C
Flash Pt: 88.6 °C
Storage Temperature: Ambient
MDL Number: MFCD03092926
CAS Number: 22282-70-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR