2-Fluoro-4-iodophenol 97%

Supplier: Ambeed
Warning

A113149-250MG A113149-1G
103798-170EA 17 USD
103798-170 103824-010 76821-966 77310-840
2-Fluoro-4-iodophenol 97%
2-Fluoro-4-iodophenol

Precaución: for Research Use Only

Formula: C₆H₄FIO
MW: 237.998 g/mol
Boiling Pt: 238.1 °C (760 mm Hg)
Flash Pt: 97.8 °C
Storage Temperature: Freezer
MDL Number: MFCD11109769
CAS Number: 2713-28-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR