2-Fluoro-4-iodonicotinic acid 95%

Supplier: Ambeed
Warning

Synonyms: 2-Fluoro-4-iodo-nicotinic acid

A219811-1G A219811-5G A219811-10G A219811-250MG
103818-818EA 9 USD
103818-818 103849-700 77088-268 77088-270 77112-708 77112-710
2-Fluoro-4-iodonicotinic acid 95%
2-Fluoro-4-iodonicotinic acid

Precaución: for Research Use Only

Formula: C₆H₃FINO₂
MW: 267.0000 g/mol
Boiling Pt: 347 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD03095310
CAS Number: 884494-51-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR