2-Fluoro-4-iodonicotinaldehyde 97%

Supplier: Ambeed
Warning

Synonyms: 2-Fluoro-4-iodopyridine-3-carboxaldehyde

A213238-1G A213238-10G
103818-272EA 11 USD
103818-272 103848-978 77087-474 77881-511
2-Fluoro-4-iodonicotinaldehyde 97%
2-Fluoro-4-iodonicotinaldehyde

Precaución: for Research Use Only

Formula: C₆H₃FINO
Boiling Pt: 304.2 °C (760 mm Hg)
Storage Temperature: Freezer
MDL Number: MFCD03095290
CAS Number: 153034-82-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR