2-Fluoro-4-iodobenzaldehyde 97%

Supplier: Ambeed
Warning

A441896-10G A441896-250MG A441896-1G
103827-680EA 35 USD
103827-680 103838-750 76774-048 76774-050 76842-520
2-Fluoro-4-iodobenzaldehyde 97%
2-Fluoro-4-iodobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄FIO
MW: 250.01 g/mol
Storage Temperature: Freezer
MDL Number: MFCD07782056
CAS Number: 699016-40-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR