2-Fluoro-4-iodo-6-methylpyridine 95%

Supplier: Ambeed
Warning

Synonyms: 2-Fluoro-4-iodo-6-picoline

A128725-250MG A128725-1G
103801-176EA 108 USD
103801-176 77026-948 77298-320
2-Fluoro-4-iodo-6-methylpyridine 95%
2-Fluoro-4-iodo-6-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₅FIN
MW: 237.02 g/mol
Boiling Pt: 230.273 °C (760 mm Hg)
Flash Pt: 93.066 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03095298
CAS Number: 884494-45-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR