2-Fluoro-4-iodo-3-methylpyridine 98%

Supplier: Ambeed
Warning

Synonyms: 2-Fluoro-4-iodo-3-picoline

A181422-5G A181422-250MG
103807-602EA 25 USD
103807-602 103845-974 77083-504 77096-870
2-Fluoro-4-iodo-3-methylpyridine 98%
2-Fluoro-4-iodo-3-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₅FIN
MW: 237.02 g/mol
Boiling Pt: 244 °C (760 mm Hg)
Flash Pt: 101 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03095289
CAS Number: 153034-80-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR