4-Cyano-3-fluorobenzaldehyde 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Fluoro-4-formylbenzonitrile, 4-Cyano-3-fluorobenzaldehyde
A106736-250MG
A106736-10G
103797-066EA
18
USD
103797-066
103822-824
77008-638
77570-406
4-Cyano-3-fluorobenzaldehyde 97%
4-Cyano-3-fluorobenzaldehyde
Precaución: for Research Use Only
Formula:
C₈H₄FNO MW: 149.1200 g/mol Melting Pt: 86…90 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD06738989 CAS Number: 101048-76-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...