α,α,α,2-Tetrafluoro-p-toluic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-Fluoro-4-(trifluoromethyl)benzoic acid

A115280-100G A115280-10G A115280-25G A115280-500G A115280-1G A115280-5G
103798-538EA 10 USD
103798-538 103824-392 77189-336 77189-338 77189-340 77189-342 77297-358 77311-202
α,α,α,2-Tetrafluoro-p-toluic acid 98%
α,α,α,2-Tetrafluoro-p-toluic acid

Precaución: for Research Use Only

Formula: C₈H₄F₄O₂
MW: 208.11 g/mol
Melting Pt: 168…170 °C
Flash Pt: 93.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00010322
CAS Number: 115029-24-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR