2-Fluoro-4-(trifluoromethyl)benzaldehyde 97%

Supplier: Ambeed
Warning

A318299-100G A318299-10G A318299-1G A318299-25G A318299-5G
103825-166EA 14 USD
103825-166 103836-098 76772-892 76772-894 76838-954 77306-174 77306-176
2-Fluoro-4-(trifluoromethyl)benzaldehyde 97%
2-Fluoro-4-(trifluoromethyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₈H₄F₄O
MW: 192.11 g/mol
Boiling Pt: 118…119 °C (760 mm Hg)
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00061310
CAS Number: 89763-93-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR