2-Fluoro-4-(trifluoromethyl)aniline 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Amino-3-fluorobenzotrifluoride, α,α,α,2-Tetrafluoro-p-toluidine, 4-Amino-alpha,alpha,alpha,3-tetrafluorotoluene, 4-Amino-α,α,α,3-tetrafluorotoluene
A343951-1G
A343951-5G
A343951-100G
A343951-10G
A343951-25G
103825-898EA
20
USD
103825-898
103836-876
76839-920
76839-922
77290-566
77290-568
77290-570
2-Fluoro-4-(trifluoromethyl)aniline 98%
2-Fluoro-4-(trifluoromethyl)aniline
Precaución: for Research Use Only
Formula:
C₇H₅F₄N MW: 179.12 Boiling Pt: 172.8 °C (760 mm Hg) Storage Temperature: Refrigerator |
MDL Number:
MFCD01631629 CAS Number: 69409-98-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...