2-Fluoro-4-pyridineboronic acid pinacol ester 97%

Supplier: Ambeed
Warning

Synonyms: 2-Fluoropyridine-4-boronic acid pinacol ester, 2-Fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

A119324-100MG A119324-10G A119324-250MG A119324-1G A119324-5G A119324-25G
103799-754EA 15 USD
103799-754 77258-042 77258-044 77258-046 77311-878 77311-880 77747-869
2-Fluoro-4-pyridineboronic acid pinacol ester 97%
2-Fluoro-4-pyridineboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₁H₁₅BFNO₂
Boiling Pt: 312.1±27.0 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD06798253
CAS Number: 458532-86-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR