3-Fluoro-4-hydroxyphenylboronic acid, pinacol ester 97%

Supplier: Ambeed
Warning

Synonyms: 2-Fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol

A132560-100MG A132560-250MG A132560-25G A132560-5G
103801-748EA 55 USD
103801-748 76823-068 76823-070 77314-332 77314-334
3-Fluoro-4-hydroxyphenylboronic acid, pinacol ester 97%
3-Fluoro-4-hydroxyphenylboronic acid, pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₆BFO₃
MW: 238.063 g/mol
Boiling Pt: 324.9 °C (760 mm Hg)
Melting Pt: 99.3…100.2 °C
Flash Pt: 150.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11504962
CAS Number: 760990-08-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR