4-Cyano-3-fluorobenzeneboronic acid pinacol ester 97%

Supplier: Ambeed
Warning

A214282-1G A214282-5G A214282-100G A214282-10G A214282-250MG A214282-25G
103818-386EA 13 USD
103818-386 103849-092 77087-604 77087-606 77152-542 77152-544 77152-546 77152-548
4-Cyano-3-fluorobenzeneboronic acid pinacol ester 97%
4-Cyano-3-fluorobenzeneboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₅BFNO₂
Boiling Pt: 346.6±32.0 °C (760 mm Hg)
Flash Pt: 163.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD16996321
CAS Number: 870238-67-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR