3-Fluoro-4-formylbenzeneboronic acid pinacol ester 95%

Supplier: Ambeed
Warning

Synonyms: 2-Fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde

A156724-10G A156724-250MG A156724-5G A156724-1G
103804-916EA 24 USD
103804-916 76823-974 76823-976 76823-978 77318-916
3-Fluoro-4-formylbenzeneboronic acid pinacol ester 95%
3-Fluoro-4-formylbenzeneboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₆BFO₃
MW: 250.076 g/mol
Boiling Pt: 343.9±32.0 °C (760 mm Hg)
Flash Pt: 161.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD16036135
CAS Number: 503176-50-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR