2-Fluoro-3-nitrobenzotrifluoride 95%

Supplier: Ambeed
Warning

A135203-10G A135203-1G A135203-25G A135203-5G
103802-110EA 27 USD
103802-110 76972-304 77314-900 77314-902 77314-904
2-Fluoro-3-nitrobenzotrifluoride 95%
2-Fluoro-3-nitrobenzotrifluoride

Precaución: for Research Use Only

Formula: C₇H₃F₄NO₂
Storage Temperature: Ambient
MDL Number: MFCD08532487
CAS Number: 61324-97-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR